Learn More
Sino Biological Recombinant Mouse TOP1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 58668M07B11
Encompass_Preferred
A DNA sequence encoding the Mouse TOP1 (NM_009408.2) (Lys193-Phe767) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-241-1114