Learn More
Biomatik Corporation Mouse Topoisomerase II (TOP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07769
Encompass_Preferred
Mouse Topoisomerase II (TOP2) ELISA Kit for detecting TOP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01320. Gene ID: 21973. Target Synonyms: DNA topoisomerase 2-alpha; DNA topoisomerase II, alpha isozyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6231