Learn More
Alpco Ltd Mouse Total Adiponectin ELISA
SDP
Supplier:  Alpco Ltd 22ADPMSE01
Encompass_Preferred
The Mouse Total Adiponectin ELISA is for the quantitative determination of total adiponectin in mouse serum, plasma, and cell culture supernatant. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2318933