Learn More
Chondrex Inc MOUSE TOTAL IGE ASSAY, 1 KIT
Catalog No. 501526734
Encompass_Preferred
ELISA kit to determine IgEa and IgEb antibody levels in mouse serum and plasma samples. This kit includes one 96-well ELISA plate with removable strips, as well as all reagents necessary to complete the assay. This kit is ideals for assaying IgE antibody levels in mouse allergy models. Standard Range: 100 ng/ml to 1.6 ng/ml. Working Time: 4.5 hours.
Catalog No. 50-152-6734 Supplier Chondrex Inc Supplier No. 3005
ELISA kit to determine IgEa and IgEb antibody levels in mouse serum and plasma samples. This kit includes one 96-well ELISA plate with removable strips, as well as all reagents necessary to complete the assay. This kit is ideals for assaying IgE antibody levels in mouse allergy models. Standard Range: 100 ng/ml to 1.6 ng/ml. Working Time: 4.5 hours.