Learn More
Chondrex Inc Mouse Total IgG Antibody Detection Kit
Supplier:  Chondrex Inc 3023
Encompass_Preferred
This mouse total immunoglobulin IgG detection ELISA kit is suitable for quantifying monoclonal antibody and polyclonal antibody isotypes, establishing reliable methods for monitoring antibody production and quality control, and eveluating a ratio of IgG to antigen-specific antibody levels to evaluate immune function in animal disease models. ASSAY TIME: 4.5 hours, STANDARD RANGE: 100 ng/ml to 1.6 ng/mlNUMBER OF SAMPLES: Up to 40 (duplicate) samples/plate, SAMPLE TYPES: Serum & Plasma (pre-treatment acceptable).
Catalog No. NC1929620