Learn More
Invitrogen™ IgG (Total) Mouse ELISA Standard
Description
Specifications
Specifications
| Format | Kit |
| Quantity | 4 pack |
| Recombinant | Recombinant |
| For Use With (Equipment) | Microplate Reader |
| Former Identifiers | eBioscience |
| Immunoassay Kit Format | ELISA Standard |
| Target Species Primary | Mouse |
| Target Kit Named | IgG |
| Target Species Named | Mouse |
| Target Species Validated | Mouse |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.