Learn More
BIOHIPPO Mouse tPA Microsample
SDP
Catalog No. 5000314007
Encompass
Mouse tPA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of tPA in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse tPA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse tPA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse tPA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14007 Supplier BIOHIPPO Supplier No. BHE15211978
Mouse tPA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of tPA in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse tPA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse tPA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse tPA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color