Learn More
ACROBiosystems ACROBiosystems Mouse TPBG / 5T4 Protein, His Tag
Catalog No. 502106900
Encompass_Preferred
Recombinant Protein;1MG;Mouse TPBG, His Tag (TPG-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 361 (Accession # Q9Z0L0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.8 kDa.
Catalog No. 50-210-6900 Supplier ACROBiosystems Supplier No. TPGM52H31MG
Recombinant Protein;1MG;Mouse TPBG, His Tag (TPG-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 361 (Accession # Q9Z0L0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.8 kDa.