Learn More
BIOHIPPO Mouse TRAF6 Microsample
SDP
Catalog No. 5000314817
Encompass
Mouse TRAF6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TRAF6 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 g/mL with a sensitivity of 90 7 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TRAF6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TRAF6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TRAF6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14817 Supplier BIOHIPPO Supplier No. BHE15212475
Mouse TRAF6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TRAF6 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 g/mL with a sensitivity of 90 7 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TRAF6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TRAF6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TRAF6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color