Learn More
Braintree Scientific Mouse Transfer Tube,Clear Polycarbonate, 130mm x 50mm dia.
SDP
Catalog No. 501956067
Encompass
Mouse TransferTube Enrichment and transfer tube, 130mm x 50mm dia. Tunnel handling is a proven method to reduce stress and anxiety when transferring a mouse. This polycarbonate tube can withstand the high temperatures of an autoclave and also maintain its shape. Please check out the tutorial for more information https://www.nc3rs.org.uk/mouse-handling-tutorial **Sold for research purposes only and not for human use**
Catalog No. 50-195-6067 Supplier Braintree Scientific Supplier No. TRANSTUBE100
Mouse TransferTube Enrichment and transfer tube, 130mm x 50mm dia. Tunnel handling is a proven method to reduce stress and anxiety when transferring a mouse. This polycarbonate tube can withstand the high temperatures of an autoclave and also maintain its shape. Please check out the tutorial for more information https://www.nc3rs.org.uk/mouse-handling-tutorial **Sold for research purposes only and not for human use**