Learn More
Rockland Immunochemicals MOUSE TRANSFERRIN - 010-0134
SDP
Supplier:  Rockland Immunochemicals 0100134
Encompass
Mouse Transferrin is a blood plasma glycoprotein which binds iron and regulates the amount of available iron in circulation. Mouse Transferrin has a high affinity for iron, but this binding is reversible. Mouse Transferrin has a molecular weight of approximately 80kDa. Mouse Transferrin also plays a role in the immune system, creating environments low in iron for which many pathogenic bacteria are unable to thrive. This transferrin is iron-poor, when not bound to iron, is know as apotransferrin or the (apo) form.
Catalog No. 50-105-7359