Learn More
ACROBiosystems Biotinylated Mouse Transferrin R, His Tag, primary amine labeling (TFR-M8249) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 763 (Accession # Q62351-1).
Supplier:  ACROBiosystems TFRM8249200UG
Encompass_Preferred
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
Catalog No. 50-223-0899