Learn More
ACROBiosystems Biotinylated Mouse Transferrin R, His Tag, primary amine labeling (TFR-M8249) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 763 (Accession # Q62351-1).
Catalog No. 502230900
Encompass_Preferred
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
This item is not returnable. View return policy
Catalog No. 50-223-0900 Supplier ACROBiosystems Supplier No. TFRM824925UG
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.