Learn More
Biomatik Corporation Mouse Transforming Growth Factor Beta 3 (TGFb3) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11264
Encompass_Preferred
Mouse Transforming Growth Factor Beta 3 (TGFb3) Mini sample ELISA Kit for detecting TGFb3 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17125. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5734