Learn More
American Research Products Inc Mouse transforming growth fact
Catalog No. 5000326060
Encompass
Mouse transforming growth factorb2 TGFb2 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26060 Supplier American Research Products Inc Supplier No. CSBE09785M
Mouse transforming growth factorb2 TGFb2 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich