Learn More
American Research Products Inc Mouse Transforming growth fact
Supplier:  American Research Products Inc CSBEL023453MO
Encompass
Mouse Transforming growth factor beta receptor type 3 (TGFBR3) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28068