Learn More
American Research Products Inc Mouse Transforming Growth fact
Catalog No. 5000329901
Encompass
Mouse Transforming Growth factor b1 TGF-b1 ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 2 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29901 Supplier American Research Products Inc Supplier No. CSBE04726M
Mouse Transforming Growth factor b1 TGF-b1 ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 2 ng/mL Sample serum plasma cell culture supernates Method Sandwich