Learn More
American Research Products Inc Mouse transforming growth fact
Supplier:  American Research Products Inc CSBE07290M
Encompass
Mouse transforming growth factor a TGF-a ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28973