Learn More
Biomatik Corporation Mouse Transglutaminase 3, Epidermal (TGM3) ELISA Kit
Supplier:  Biomatik Corporation EKU07834
Encompass_Preferred
Mouse Transglutaminase 3, Epidermal (TGM3) ELISA Kit for detecting TGM3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.138ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08189. Gene ID: 21818. Target Synonyms: TGE; Egt; Protein-Glutamine Gamma-Glutamyltransferase E; Transglutaminase E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7227