Learn More
Biomatik Corporation Mouse Transient Receptor Potential Cation Channel Subfamily V, Member 2 (TRPV2) ELISA Kit
Supplier:  Biomatik Corporation EKU07846
Encompass_Preferred
Mouse Transient Receptor Potential Cation Channel Subfamily V, Member 2 (TRPV2) ELISA Kit for detecting TRPV2 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTR1. Gene ID: 22368. Target Synonyms: VRL; OTRPC2; VRL1; Osm-9-like TRP channel 2; Vanilloid receptor-like protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7423