Learn More
Biomatik Corporation Mouse Translocation Associated Notch Homolog 1 (TAN1) ELISA Kit
Supplier:  Biomatik Corporation EKU10247
Encompass_Preferred
Mouse Translocation Associated Notch Homolog 1 (TAN1) ELISA Kit for detecting TAN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01705. Gene ID: 18128. Target Synonyms: NOTCH1; hN1; NEXT; NICD; Translocation-associated notch protein TAN-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4630