Learn More
American Research Products Inc Mouse Transmembrane glycoprote
Catalog No. 5000327827
Encompass
Mouse Transmembrane glycoprotein NMB (GPNMB) ELISA kit from CUSABIO Detection Range 0 78 ng/ml-50 ng/ml Sensitivity 0 195ng/ml Sample serum plasma tissue homogenates Method
Catalog No. 50-003-27827 Supplier American Research Products Inc Supplier No. CSBEL009727MO
Mouse Transmembrane glycoprotein NMB (GPNMB) ELISA kit from CUSABIO Detection Range 0 78 ng/ml-50 ng/ml Sensitivity 0 195ng/ml Sample serum plasma tissue homogenates Method