Learn More
Biomatik Corporation Mouse Transmembrane Protein 27 (TMEM27) ELISA Kit
Supplier:  Biomatik Corporation EKU07861
Encompass_Preferred
Mouse Transmembrane Protein 27 (TMEM27) ELISA Kit for detecting TMEM27 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESG4. Gene ID: 57394. Target Synonyms: NX17; Collectrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6809