Learn More
World Precision Instrument Mouse Transverse Clamps, Set of 4
SDP
Supplier:  World Precision Instrument 505290
Encompass
The design was inspired by the need for a practical, versatile, and economical instrument for stabilizing the spinal column. The Spinal Adaptors are accessories to be mounted on the 505211, 505351 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.
Catalog No. 50-253-9135