Learn More
Biolegend FITC anti-mouse TCR β chain
Supplier:  Biolegend 109206
Encompass_Preferred
FITC anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F; Size: 500 ug
Catalog No. NC1455814