Learn More
Sino Biological Anti-TREM-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50149RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TREM-2 / TREM2 (rM TREM-2 / TREM2; Catalog#50149-M08H; NP_112544.1; Met1-Pro168). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2479