Learn More
Sino Biological Recombinant Mouse TREM-2 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50149M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse TREM2 (NP_112544.1)(Met1-Ser171) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-2023