Learn More
Sino Biological Recombinant Mouse TRIM21 Protein (His & AVI Tag), Avi-tag Biotinylated
Catalog No. 502992735
Encompass_Preferred
A DNA sequence encoding the Mouse TRIM21 (NP_001076021.1) (Met1-Met462) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2735 Supplier Sino Biological Supplier No. 59826M56BB100
A DNA sequence encoding the Mouse TRIM21 (NP_001076021.1) (Met1-Met462) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.