Learn More
ACROBiosystems Mouse TROP-2 25ug
Catalog No. 502534015
Encompass_Preferred
Biotinylated Mouse TROP-2 Protein FcAvitag (TR2-M82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gly 270 (Accession # Q8BGV3-1).
Catalog No. 50-253-4015 Supplier ACROBiosystems Supplier No. TR2M82F325UG
Biotinylated Mouse TROP-2 Protein FcAvitag (TR2-M82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gly 270 (Accession # Q8BGV3-1).