Learn More
Sino Biological Recombinant Mouse TROP-2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50922M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TACSTD2 (Q8BGV3) (Met1-Gly270) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-6850