Learn More
Biomatik Corporation Mouse Tropomyosin 3 (TPM3) ELISA Kit
Supplier:  Biomatik Corporation EKU08483
Encompass_Preferred
Mouse Tropomyosin 3 (TPM3) ELISA Kit for detecting TPM3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21107. Gene ID: 59069. Target Synonyms: NEM1; TRK; OK/SW-cl.5; TM-5; TM3; TM30; TM30nm; TPMsk3; Hscp30; Gamma-tropomyosin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6727