Learn More
American Research Products Inc Mouse Troponin T cardiac musc
Catalog No. 5000327212
Encompass
Mouse Troponin T cardiac muscle (TNNT2) ELISA kit from CUSABIO Detection Range 31 25 pg/ml-2000pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27212 Supplier American Research Products Inc Supplier No. CSBEL024016MO
Mouse Troponin T cardiac muscle (TNNT2) ELISA kit from CUSABIO Detection Range 31 25 pg/ml-2000pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich