Learn More
Rockland Immunochemicals Mouse TrueBlot Set (with IP Agarose beads) - 88-7788-31
SDP
Catalog No. 501058851
Encompass_Preferred
Mouse IgG TrueBlot ULTRA is provided as 1000X solution. We recommend incubating the blots from minigels in sealed bags with minimal volume (2-3 mLs). If used conservatively at 2.5mls/blot will yield enough reagent for 20 blots. Note that there are three key procedural considerations: 1. Protein A or G beads may be used with the mouse, goat and sheep TrueBlot secondaries, but not with the rabbit TrueBlot secondary. Use of protein A or G beads with the rabbit TrueBlot will result in contaminating bands. 2. Immunoprecipitate should be completely reduced. 3. BLOTTO/Milk should be used as the blocking protein for the immunoblot. We recommend using a large bore pipet to pipet up the liquid for use. To achieve best results when detecting mouse IgG1 subtypes, we recommend performing a dot blot or titration to determine the optimal dilution. Specific conditions for each protein and antibody combination should be specifically optimized by the end user.
Catalog No. 50-105-8851 Supplier Rockland Immunochemicals Supplier No. 88778831
Mouse IgG TrueBlot ULTRA is provided as 1000X solution. We recommend incubating the blots from minigels in sealed bags with minimal volume (2-3 mLs). If used conservatively at 2.5mls/blot will yield enough reagent for 20 blots. Note that there are three key procedural considerations: 1. Protein A or G beads may be used with the mouse, goat and sheep TrueBlot secondaries, but not with the rabbit TrueBlot secondary. Use of protein A or G beads with the rabbit TrueBlot will result in contaminating bands. 2. Immunoprecipitate should be completely reduced. 3. BLOTTO/Milk should be used as the blocking protein for the immunoblot. We recommend using a large bore pipet to pipet up the liquid for use. To achieve best results when detecting mouse IgG1 subtypes, we recommend performing a dot blot or titration to determine the optimal dilution. Specific conditions for each protein and antibody combination should be specifically optimized by the end user.