Learn More
Rockland Immunochemicals Mouse TrueBlot ULTRA: Anti-Mouse Ig HRP - 18-8817-30
SDP
Supplier:  Rockland Immunochemicals 18881730
Encompass_Preferred
Mouse TrueBlot ULTRA has been reported for use in IP and WB. Mouse IgG TrueBlot ULTRA is provided as 1000X solution. To achieve best results when detecting mouse IgG1 subtypes, we recommend performing a dot blot or titration to determine the optimal dilution. Specific conditions for each protein and antibody combination should be specifically optimized by the end user. We recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mLs). If used conservatively at 2.5mls/blot will yield enough reagent for 80 blots. Note that there are three key procedural considerations: 1. Protein A or G beads may be used with the mouse, goat and sheep TrueBlot secondaries, but not with the rabbit TrueBlot secondary. Use of protein A or G beads with the rabbit TrueBlot will result in contaminating bands. 2. Immunoprecipitate should be completely reduced. 3. BLOTTO/Milk should be used as the blocking protein for the immunoblot.
Catalog No. NC0412690