Learn More
American Research Products Inc Mouse Tryptase beta 2 TPSB2
Catalog No. 5000327902
Encompass
Mouse Tryptase beta-2 (TPSB2) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27902 Supplier American Research Products Inc Supplier No. CSBEL024128MO
Mouse Tryptase beta-2 (TPSB2) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich