Learn More
Sino Biological Recombinant Mouse TSLP Protein (His Tag)
Supplier:  Sino Biological 51005M08H100
Encompass_Preferred
A DNA sequence encoding the mouse TSLP (Q9JIE6) (Met 1-Glu 140) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-228-2822