Learn More
Sino Biological Mouse Tssk3 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 5A1305M09B100
Encompass_Preferred
A DNA sequence encoding the Mouse Tssk3 (NP_536690.1) (Met1-Thr268) was expressed with an N-terminal GST tag.
Catalog No. 50-237-3674