Learn More
Biomatik Corporation Mouse Tubulin Beta 1 (TUBb1) ELISA Kit
Supplier:  Biomatik Corporation EKU07936
Encompass_Preferred
Mouse Tubulin Beta 1 (TUBb1) ELISA Kit for detecting TUBb1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A2AQ07. Gene ID: 545486. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8712