Learn More
Sigma Aldrich Fine Chemicals Biosciences Mouse Tumor Necrosis facto
Catalog No. 502906600
Encompass
Tumor Necrosis factor (TNf ) is a proinflammatory cytokine. It is a transmembrane protein and the final protein is devoid of the extracellular domain. TNf -converting enzyme (TACE) catalyzes the formation of the soluble homotrimer TNf .
Catalog No. 50-290-6600 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB04771KT
Tumor Necrosis factor (TNf ) is a proinflammatory cytokine. It is a transmembrane protein and the final protein is devoid of the extracellular domain. TNf -converting enzyme (TACE) catalyzes the formation of the soluble homotrimer TNf .