Learn More
American Research Products Inc Mouse Tumor necrosis factor a
Catalog No. 5000328774
Encompass
Mouse Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28774 Supplier American Research Products Inc Supplier No. CSBE04741M1
Mouse Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample cell culture supernates tissue homogenates Method Sandwich