Learn More
American Research Products Inc Mouse Tumor necrosis factor a
Catalog No. 5000329121
Encompass
Mouse Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/ml Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-29121 Supplier American Research Products Inc Supplier No. CSBE04741MIS
Mouse Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/ml Sample serum plasma tissue homogenates cell culture supernates Method Sandwich