Learn More
American Research Products Inc Mouse Tumor necrosis factor a
Catalog No. 5000329377
Encompass
Mouse Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29377 Supplier American Research Products Inc Supplier No. CSBE04741M
Mouse Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich