Learn More
American Research Products Inc Mouse tumor necrosis factor a
Catalog No. 5000327243
Encompass
Mouse tumor necrosis factor a (TNF-a) antibody ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27243 Supplier American Research Products Inc Supplier No. CSBEQ023955MO
Mouse tumor necrosis factor a (TNF-a) antibody ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma Method Sandwich