Learn More
American Research Products Inc Mouse Tumor necrosis factor b
Catalog No. 5000326310
Encompass
Mouse Tumor necrosis factor b TNF-b ELISA KIT from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26310 Supplier American Research Products Inc Supplier No. CSBE04744M
Mouse Tumor necrosis factor b TNF-b ELISA KIT from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich