Learn More
American Research Products Inc Mouse Tumor necrosis factor in
Catalog No. 5000327680
Encompass
Mouse Tumor necrosis factor-inducible gene 6 protein (TNFAIP6) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27680 Supplier American Research Products Inc Supplier No. CSBEL023959MO
Mouse Tumor necrosis factor-inducible gene 6 protein (TNFAIP6) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich