Learn More
American Research Products Inc Mouse Tumor necrosis factor li
Supplier:  American Research Products Inc CSBEL023989MO
Encompass
Mouse Tumor necrosis factor ligand superfamily member 13 (TNFSF13) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 43 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27682