Learn More
American Research Products Inc Mouse Tumor necrosis factor li
Supplier:  American Research Products Inc CSBEL023987MO
Encompass
Mouse Tumor necrosis factor ligand superfamily member 12 (TNFSF12) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27814