Learn More
American Research Products Inc Mouse Tumor necrosis factor li
Catalog No. 5000328071
Encompass
Mouse Tumor necrosis factor ligand superfamily member 14 (TNFSF14) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28071 Supplier American Research Products Inc Supplier No. CSBEL023991MO
Mouse Tumor necrosis factor ligand superfamily member 14 (TNFSF14) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich