Learn More
American Research Products Inc Mouse Tumor necrosis factor li
Supplier:  American Research Products Inc CSBEL023997MO
Encompass
Mouse Tumor necrosis factor ligand superfamily member 9 (TNFSF9) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27801