Learn More
American Research Products Inc Mouse Tumor necrosis factor li
Supplier:  American Research Products Inc CSBEL023994MO
Encompass
Mouse Tumor necrosis factor ligand superfamily member 4 (TNFSF4) ELISA kit from CUSABIO Detection Range 15 6pg/mL-1000pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28262